Learn More
MSC Sontara EC Engineered-Cloth Wipers, 12 in x 12 in, Creped, Flat, White
Catalog No. 502838195
Encompass
These engineered-cloth wipers are designed for critical environments, offering low lint and alcohol-free properties. They feature a flat sheet, dry style with a light dry crepe construction, ideal for cleanroom applications and delicate task preparation. Each sheet measures 13 in by 12 in and is white in color.
  • 100 sheets per box, 10 boxes per case
  • Low lint for critical environments
  • Alcohol-free composition
  • Flat sheet form for versatile use
  • Dry style for controlled application
  • Light dry crepe construction
  • White color for easy visibility
  • Suitable for aerospace, education, and pharmaceutical applications
  • Ideal for cleanroom and delicate task applications
  • Effective for surface preparation before critical tasks
Catalog No. 50-283-8195 Supplier MSC Supplier No. 37477460
These engineered-cloth wipers are designed for critical environments, offering low lint and alcohol-free properties. They feature a flat sheet, dry style with a light dry crepe construction, ideal for cleanroom applications and delicate task preparation. Each sheet measures 13 in by 12 in and is white in color.
  • 100 sheets per box, 10 boxes per case
  • Low lint for critical environments
  • Alcohol-free composition
  • Flat sheet form for versatile use
  • Dry style for controlled application
  • Light dry crepe construction
  • White color for easy visibility
  • Suitable for aerospace, education, and pharmaceutical applications
  • Ideal for cleanroom and delicate task applications
  • Effective for surface preparation before critical tasks