Learn More
MSC Monarch Brands Spunlace Interfold Wiper, Heavy Duty, Blue
Catalog No. 502838260
Encompass
These advanced performance wipers are designed as high-performance non-woven cloth and shop towel replacements. They are heavy-duty, dry, and creped spunlace interfold wipers, ideal for industrial and automotive environments. They offer solvent-resistance, virtually lint-free wiping, and exceptional durability.
  • Highly absorbent for spills, grease, and oils
  • High tensile strength, even when wet, for aggressive wiping
  • Durable and resistant to a broad range of solvents
  • Virtually lint-free, crucial for sensitive applications
  • Fragrance and alcohol-free
  • Creped and heavy-duty construction
  • Cost-effective replacement for traditional shop towels and rags
Catalog No. 50-283-8260 Supplier MSC Supplier No. 37477361
These advanced performance wipers are designed as high-performance non-woven cloth and shop towel replacements. They are heavy-duty, dry, and creped spunlace interfold wipers, ideal for industrial and automotive environments. They offer solvent-resistance, virtually lint-free wiping, and exceptional durability.
  • Highly absorbent for spills, grease, and oils
  • High tensile strength, even when wet, for aggressive wiping
  • Durable and resistant to a broad range of solvents
  • Virtually lint-free, crucial for sensitive applications
  • Fragrance and alcohol-free
  • Creped and heavy-duty construction
  • Cost-effective replacement for traditional shop towels and rags