Learn More
Perkin Elmer US LLC Stainless Steel High Pressure Piston for Flexar LC or Series 200
Catalog No. 509044063
Encompass_Preferred
Check valves, which include a sealing washer, are easy to install using the optional Torque Wrench kit. For all PerkinElmer pumps an intermediate check valve is required. The input check valve, which is identical, should be ordered.
Catalog No. 50-904-4063 Supplier Perkin Elmer US LLC Supplier No. N2600124
Check valves, which include a sealing washer, are easy to install using the optional Torque Wrench kit. For all PerkinElmer pumps an intermediate check valve is required. The input check valve, which is identical, should be ordered.