Learn More
AVANTOR MASTERFLEX (VWR LLC) High Pressure tubing with Luer Ends, 900 PSI, 20"", Non-sterile
Catalog No. NC0982407
Encompass
High Pressure tubing with Luer Ends, 900 PSI, 20", Non-sterile, High pressure tubing has luer fittings at each end¿ male luer lok x female luer in varying lengths from 10" to 48". Can be sterilized by ETO.Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Catalog No. NC0982407 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3052616
High Pressure tubing with Luer Ends, 900 PSI, 20", Non-sterile, High pressure tubing has luer fittings at each end¿ male luer lok x female luer in varying lengths from 10" to 48". Can be sterilized by ETO.Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications