Learn More
Cole-Parmer HI-PURITY SILICONE STOPPER SZ2
Supplier:  Cole-Parmer 6299414
Encompass_Preferred
High-purity silicone stopper, size 2 (21D)Stop particle buildupConstructed of high-purity silicone to resist particle build-upTranslucentAutoclavable
Catalog No. NC2471522