Learn More
Hi Q Environmental Products HI-Q TC-12 Cartridge, TEDA Impregnated Carbon, 10 Cartridges
SDP
Catalog No. NC3725204
Encompass
The HI-Q Environmental Products TC-12 offers an efficient solution for iodine species adsorption through its TEDA impregnated carbon design. This cartridge is engineered for compatibility with most industry-standard holders, ensuring ease of use and analysis. Constructed from high-density polypropylene, it also boasts a ten-year shelf life due to its heat-sealed airtight packaging. This durability and reliability differentiate it from other cartridges in the market, making it a valuable choice for those requiring high-performance filtration.
Catalog No. NC3725204 Supplier Hi Q Environmental Products Supplier No. TC12
The HI-Q Environmental Products TC-12 offers an efficient solution for iodine species adsorption through its TEDA impregnated carbon design. This cartridge is engineered for compatibility with most industry-standard holders, ensuring ease of use and analysis. Constructed from high-density polypropylene, it also boasts a ten-year shelf life due to its heat-sealed airtight packaging. This durability and reliability differentiate it from other cartridges in the market, making it a valuable choice for those requiring high-performance filtration.