Learn More
Agilent Technologies High Sensitivity D1000 ScreenTape for DNA Analysis
Catalog No. NC1786959
Encompass
High Sensitivity D1000 ScreenTape. For the high sensitivity analysis of DNA from 35 to 1000 bp. Includes 7 ScreenTape devices.
Catalog No. NC1786959 Supplier Agilent Technologies Supplier No. 50675584
High Sensitivity D1000 ScreenTape. For the high sensitivity analysis of DNA from 35 to 1000 bp. Includes 7 ScreenTape devices.