Learn More
Agilent Technologies High Sensitivity Protein 250 Ladder, 10-250 kDa, 100 Samples/Kit
Catalog No. NC1846973
Encompass
The High Sensitivity Protein 250 Ladder is designed for protein sizing within the range of 10 kDa to 250 kDa. This reagent kit is manufactured under ISO 9001:2015 quality processes and is intended exclusively for research applications. The ladder is used in conjunction with labeling dyes and provides sufficient material for three independent labeling reactions, each capable of being used for multiple chips.
  • Sizing range of 10 kDa to 250 kDa
  • Sizing resolution of 10 %
  • Typical sizing accuracy of ±10 % CV (BSA)
  • Sizing reproducibility of 3 % CV
  • Sensitivity of 1 pg/μL labeled BSA in water on chip
  • Quantitative range of 0.3 to 3,000 ng/L BSA
  • Designed for 10 chips with a 5 μL sample volume
  • Analysis time of 30 min
  • Minimum shelf life of 6 months when stored at -20°C
Catalog No. NC1846973 Supplier Agilent Technologies Supplier No. 50671578
The High Sensitivity Protein 250 Ladder is designed for protein sizing within the range of 10 kDa to 250 kDa. This reagent kit is manufactured under ISO 9001:2015 quality processes and is intended exclusively for research applications. The ladder is used in conjunction with labeling dyes and provides sufficient material for three independent labeling reactions, each capable of being used for multiple chips.
  • Sizing range of 10 kDa to 250 kDa
  • Sizing resolution of 10 %
  • Typical sizing accuracy of ±10 % CV (BSA)
  • Sizing reproducibility of 3 % CV
  • Sensitivity of 1 pg/μL labeled BSA in water on chip
  • Quantitative range of 0.3 to 3,000 ng/L BSA
  • Designed for 10 chips with a 5 μL sample volume
  • Analysis time of 30 min
  • Minimum shelf life of 6 months when stored at -20°C