Learn More
Agilent Technologies Agilent RNA 6000 Pico Ladder
Catalog No. NC1639348
Encompass
Agilent RNA 6000 Pico Ladder. Ladder only. For Bioanalyzer High Sensitivity RNA Analysis
Catalog No. NC1639348 Supplier Agilent Technologies Supplier No. 50671535
Agilent RNA 6000 Pico Ladder. Ladder only. For Bioanalyzer High Sensitivity RNA Analysis