Learn More
IBI SCIENTIFIC 3P Hi-Speed Mini Plasmid Kit, 100 preps
Catalog No. NC0753503
Encompass_Preferred
High-Speed Plasmid Mini Kit - 100 preps Sample: 1-4ml of bacterial culture Format: Spin column Operation: Centrifuge or vaccum manifold Binding Capacity: Up to 30ug Expected Yield: 20-30ug for high-copy plasmid 3-10ug for low-copy plasmid.
Catalog No. NC0753503 Supplier IBI SCIENTIFIC 3P Supplier No. IB47101
High-Speed Plasmid Mini Kit - 100 preps Sample: 1-4ml of bacterial culture Format: Spin column Operation: Centrifuge or vaccum manifold Binding Capacity: Up to 30ug Expected Yield: 20-30ug for high-copy plasmid 3-10ug for low-copy plasmid.