Learn More
iQ Biosciences Human Induced T Regulatory Cells, 1M cells/vial
Catalog No. NC2313945
Encompass_Preferred
High-quality, high-viability (typically > 80%) induced/adaptive human T regulatory cells generated from peripheral CD4 cells with confirmed suppressive capacity. Used for a wide variety of in-vitro applications, including co-culture suppression and cytotoxicity assays. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2313945 Supplier iQ Biosciences Supplier No. IQBHU1ITR1
High-quality, high-viability (typically > 80%) induced/adaptive human T regulatory cells generated from peripheral CD4 cells with confirmed suppressive capacity. Used for a wide variety of in-vitro applications, including co-culture suppression and cytotoxicity assays. All orders come with an iQ Certificate of Analysis.