Learn More
New England Biolabs, Inc. Hi-T7 RNA Polymerase – 5000 units
SDP
Supplier:  New England Biolabs, Inc. M0658S
Encompass
Hi-T7 RNA Polymerase is an engineered DNA-dependent RNA polymerase that is highly specific for T7 phage promoters and is designed to function at higher temperatures than the wild-type bacteriophage T7 RNA Polymerase. Hi-T7 RNA Polymerase functions at an optimal temperature of 50-52°C.
Catalog No. NC1690089