Learn More
MSC HI-TEMP CREPE MASK TAPE
Catalog No. 5000686828
Encompass
This masking tape features a rubber adhesive for strong tack and adhesion, designed for clean removal from various surfaces. It is suitable for multi-bake or high-temperature bake processes, with a smooth mini-crepe paper backing that conforms well and maintains integrity.
  • Strong backing resists slivering and provides one-piece removal
  • Controlled unwind mechanism reduces user fatigue
  • Solvent-resistant and moisture-resistant properties
  • Easy tear and instant adhesion
  • Meets ASTM D-6123 standards
  • Best removal within 24 hours
Catalog No. 50-006-86828 Supplier MSC Supplier No. 02999548
This masking tape features a rubber adhesive for strong tack and adhesion, designed for clean removal from various surfaces. It is suitable for multi-bake or high-temperature bake processes, with a smooth mini-crepe paper backing that conforms well and maintains integrity.
  • Strong backing resists slivering and provides one-piece removal
  • Controlled unwind mechanism reduces user fatigue
  • Solvent-resistant and moisture-resistant properties
  • Easy tear and instant adhesion
  • Meets ASTM D-6123 standards
  • Best removal within 24 hours