Learn More
Electron Microscopy Sciences Hi-Temp Indicator Tape, 1/2" Wide 250 IN
SDP
Catalog No. 5031297
Encompass_Preferred
Specialty cloth-textured Hi Temperature Indicator tape shows the word "STERILIZED" in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550 degree F dry oven temperatures.
Catalog No. 50-312-97 Supplier Electron Microscopy Sciences Supplier No. 7701505
Specialty cloth-textured Hi Temperature Indicator tape shows the word "STERILIZED" in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550 degree F dry oven temperatures.