Learn More
Bellco Glass, Inc. Hi-Temp Red Pouring Ring,32mm
SDP
Catalog No. 501945821
Encompass_Preferred
Use with Hi-Temperature Red Cap. Pouring ring provides drip free pouring an protects thread from chipping. They are autoclavable to 180C. Packed 10 per case. MADE IN THE USA
Catalog No. 50-194-5821 Supplier Bellco Glass, Inc. Supplier No. 563791032
Use with Hi-Temperature Red Cap. Pouring ring provides drip free pouring an protects thread from chipping. They are autoclavable to 180C. Packed 10 per case. MADE IN THE USA