Learn More
Bellco Glass, Inc. Hi-Temp Red Screw Cap,32mm
SDP
Catalog No. 501945819
Encompass_Preferred
Manufactured from red Polybutylene Terephthalate with PTFE faced silicone liner. They can withstand temperature of -45 degree Celsius to 200 degrees Celsius. MADE IN THE USASize: 32mm
Catalog No. 50-194-5819 Supplier Bellco Glass, Inc. Supplier No. 563790032
Manufactured from red Polybutylene Terephthalate with PTFE faced silicone liner. They can withstand temperature of -45 degree Celsius to 200 degrees Celsius. MADE IN THE USASize: 32mm