Learn More
Bellco Glass, Inc. Hi-Temp Red Screw Cap,45mm
SDP
Catalog No. 501945820
Encompass_Preferred
Manufactured from red Polybutylene Terephthalate with PTFE faced silicone liner. They can withstand temperature of -45 degree Celsius to 200 degrees Celsius. MADE IN THE USA Size: 45mm
Catalog No. 50-194-5820 Supplier Bellco Glass, Inc. Supplier No. 563790045
Manufactured from red Polybutylene Terephthalate with PTFE faced silicone liner. They can withstand temperature of -45 degree Celsius to 200 degrees Celsius. MADE IN THE USA Size: 45mm