Learn More
Chemglass Life Sciences High Temperature Cap, GL-45, Solid, PBTE
SDP
Catalog No. 501215151
Encompass_Preferred
High temperature PBTE cap will withstand temperature from -45 to 200 degreesC. complete with PTFE seal.
Catalog No. 50-121-5151 Supplier Chemglass Life Sciences Supplier No. CLS148602
High temperature PBTE cap will withstand temperature from -45 to 200 degreesC. complete with PTFE seal.