Learn More
Clearco Products Co Inc High Temperature Silicone Bath Fluid, 1 Gallon
Catalog No. NC1414409
Encompass
DPDM-400 HIGH TEMPERATURE BATH FLUID is a clear, colorless and odorless Diphenyl-Dimethylsiloxane (CAS# 68083-14-7) with a viscosity of 400cSt. It remains stable at 250°C (open system) for 1,500 to 2,000 hours without gelling or breaking down. In closed system baths*, DPDM-400 HIGH TEMPERATURE BATH FLUID remains stable at 300°C.
Catalog No. NC1414409 Supplier Clearco Products Co Inc Supplier No. DPDM4001GAL
DPDM-400 HIGH TEMPERATURE BATH FLUID is a clear, colorless and odorless Diphenyl-Dimethylsiloxane (CAS# 68083-14-7) with a viscosity of 400cSt. It remains stable at 250°C (open system) for 1,500 to 2,000 hours without gelling or breaking down. In closed system baths*, DPDM-400 HIGH TEMPERATURE BATH FLUID remains stable at 300°C.