Learn More
Brookfield Engineer Labs HI TEMP VISCOSITY STD 500ML
Catalog No. NC9300025
Encompass
100000 CPS High Temperature Silicone Viscosity Standard
Catalog No. NC9300025 Supplier Brookfield Engineer Labs Supplier No. HT100000
100000 CPS High Temperature Silicone Viscosity Standard