Learn More
Meridian Life Science Hi-throughput dUTP qPCR Mix, 1ML
SDP
Catalog No. 5000856284
Encompass_Preferred
Hi-throughput dUTP qPCR Mix comprises a master mix containing Taq polymerase, reaction buffer, MgCl2 and optimized dNTP/dUTP mix, and PCR enhancers. It is designed for stringent amplification of highly-structured and very low copy-number DNA targets, preventing carry-over qPCR contamination, makes Hi-throughput dUTP qPCR ideal for multiplex assays on open, automated, high-throughput instruments.
Catalog No. 50-008-56284 Supplier Meridian Life Science Supplier No. MDX0321
Hi-throughput dUTP qPCR Mix comprises a master mix containing Taq polymerase, reaction buffer, MgCl2 and optimized dNTP/dUTP mix, and PCR enhancers. It is designed for stringent amplification of highly-structured and very low copy-number DNA targets, preventing carry-over qPCR contamination, makes Hi-throughput dUTP qPCR ideal for multiplex assays on open, automated, high-throughput instruments.