Learn More
Bellco Glass, Inc. Hi-Tor CTII Digital Over Head Drive Complete, 115V
SDP
Catalog No. 501946218
Encompass_Preferred
The Bellco Digital CTII Hi-Torque Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixing and the stirring of solutions with a viscosity greater than water. Hi-Torque Overhead Drive with interface for use with CellTrol II controller. Having speed capability user can monitor (but not control) rpm of OHD from the controller display and set alarm for rpm setpoints. Includes speed cable and power supply.. MADE IN THE USA
Catalog No. 50-194-6218 Supplier Bellco Glass, Inc. Supplier No. 777460115
The Bellco Digital CTII Hi-Torque Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixing and the stirring of solutions with a viscosity greater than water. Hi-Torque Overhead Drive with interface for use with CellTrol II controller. Having speed capability user can monitor (but not control) rpm of OHD from the controller display and set alarm for rpm setpoints. Includes speed cable and power supply.. MADE IN THE USA