Learn More
Bellco Glass, Inc. Hi-Torq Over Head Drive Adapter,100mm
SDP
Catalog No. 501946199
Encompass_Preferred
Hi -Torque Over Head Drive Adapter is used in conjunction with the High-Torque Over Head Drive Motor to drive the impeller assembly for mixing within a vessel. Adapts all Bellco PTFE impeller assys or SST impeller assys. Used with 100mm cap assembly. 316 SST design with polyester/SST bearing. Includes two (2) 1/4"npt female plugged ports for optional fittings.NOT HIGH SPEED!SPEED RANGE: 5- 150rpmDimensions: 3 3/4" l x 3 3/4' w x 7 1/2" tall (including dust shield coupling. MADE IN THE USA
Catalog No. 50-194-6199 Supplier Bellco Glass, Inc. Supplier No. 776474000
Hi -Torque Over Head Drive Adapter is used in conjunction with the High-Torque Over Head Drive Motor to drive the impeller assembly for mixing within a vessel. Adapts all Bellco PTFE impeller assys or SST impeller assys. Used with 100mm cap assembly. 316 SST design with polyester/SST bearing. Includes two (2) 1/4"npt female plugged ports for optional fittings.NOT HIGH SPEED!SPEED RANGE: 5- 150rpmDimensions: 3 3/4" l x 3 3/4' w x 7 1/2" tall (including dust shield coupling. MADE IN THE USA