Learn More
Ace Glass, Inc. Dow Corning, High vacuum grease, tube, 5.3 oz. (150g)
SDP
Catalog No. 509558411
Encompass
Dow Corning High Vacuum Grease. A silicone lubricant that effectively seals and lubricates glass stopcocks, joints and glass-rubber connections. Resistant to most chemicals, heat stable and inert. Supplied in tube, 5.3 oz. (150g).
Catalog No. 50-955-8411 Supplier Ace Glass, Inc. Supplier No. 823006
Dow Corning High Vacuum Grease. A silicone lubricant that effectively seals and lubricates glass stopcocks, joints and glass-rubber connections. Resistant to most chemicals, heat stable and inert. Supplied in tube, 5.3 oz. (150g).