Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Y9236 Cool Series Nitrile-Coated Hi-Vis Gloves

Catalog No. 17200355 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-355 X-Small
17-200-356 Small
17-200-357 Medium
17-200-358 Large
17-200-359 X-Large
17-200-360 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-200-355 Supplier Wells Lamont™ Supplier No. Y9236XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech™ Y9236 Cool Series Nitrile-Coated Hi-Vis Gloves feature a blend of high performance and cooling fibers that provide a high level of protection and comfort in warmer environments. Hi-vis shell improves visibility, while nitrile coating on the palm improves grip capabilities.

  • Hi-vis, cut-resistant shell improves visibility and ensures workers' compliance
  • Reinforced thumb crotch with aramid fiber stitching
  • Knit wrist prevents dirt and debris from entering the glove
  • 15-gauge seamless knit construction
  • Blend of high performance and cooling fibers
  • Sandy nitrile palm coating improves grip capabilities

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ASTM F2992-15, ASTM D3389-10
Construction Seamless Knit
Coating Material Nitrile
Color Hi-vis Yellow
Cut Resistance Rating ANSI A4
Exterior Finish Fully Textured
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size X-Small
EN388 Standard EN 388:4543
For Use With (Application) Construction, metal fabrication, assembly
Glove Cuff Color Yellow
Glove Cuff Style Knit
Grip Design Sandy nitrile
Model Y9236XS
Series FlexTech™ Y9236
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.