Learn More
Wells Lamont™ FlexTech™ Y9236 Cool Series Nitrile-Coated Hi-Vis Gloves
Shop All Wells Lamont Industry Group ProductsDescription
- Hi-vis, cut-resistant shell improves visibility and ensures workers' compliance
- Reinforced thumb crotch with aramid fiber stitching
- Knit wrist prevents dirt and debris from entering the glove
- 15-gauge seamless knit construction
- Blend of high performance and cooling fibers
- Sandy nitrile palm coating improves grip capabilities
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance | ASTM F2992-15, ASTM D3389-10 |
| Construction | Seamless Knit |
| Coating Material | Nitrile |
| Color | Hi-vis Yellow |
| Cut Resistance Rating | ANSI A4 |
| Exterior Finish | Fully Textured |
| Hand | Specific |
| Latex Free | Yes |
| Material | Aramid Fiber |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.