Learn More
Pyramex™ Hi-Vis Mesh Safety Vests
Catalog No. 19555483 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-483 3X-Large
19-555-481 Small
19-555-480 Medium
19-555-479 Large
19-555-482 2X-Large
19-555-484 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-555-483 Supplier Pyramex™ Supplier No. RVHLM2910X3
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Hi-Vis Mesh Safety Vests with ANSI/ISEA 107-2015 safety compliance
  • Front: Hook & Loop closure
  • One 2 in. silver reflective stripe applied horizontally 8.5 in. from the bottom
  • Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over shoulders to top of back side horizontal stripe
Specifications
Closure Type Hook and Loop
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 2
Color Hi-vis Lime
Size 3X-Large
Material Polyester Mesh
Size (English) Chest 54 to 56 in.
Tape Type China Brand Flat Silver