Learn More
MCR Safety™ Hi-Vis Reflective Lime Mesh Safety Vest, 5 pt. break, CSA Cl. 2
Catalog No. 19034366 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-034-366 Large
19-034-365 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-366 Supplier MCR Safety™ Supplier No. RXCL2MLL
Luminator™ Series Hi-Vis reflective lime safety vest - manufactured with recycled materials, meets ANSI/ISEA 107 2020 standard, type R class 2 compliant, five point break away design.
  • Meets ANSI/ISEA 107 2020 Standard - Type R, Class 2 compliant
  • 5 Point break away pattern
  • CSA Z96 Class 2 Level 2 compliant
  • D-Ring access slot on back
  • Hook and Loop front closure
  • Fluorescent lime mesh polyester material
  • 1 outer chest pocket
  • Made of recycled materials
  • 2 Inch silver reflective stripes
  • X reflective pattern on vest back
Specifications
Product Type High-visibility Vest
Closure Type Breakaway Hook and Loop
Color Hi-vis Lime
Size Large
Material Polyester Mesh
No. of Pockets 1