Learn More
Pyramex™ Hi-Vis Reflective Safety Vests
Catalog No. 19555469 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-469 Hi-vis Lime Medium
19-555-468 Hi-vis Lime Large
19-555-473 Hi-vis Lime X-Large
19-555-470 Hi-vis Lime Small
19-555-471 Hi-vis Lime 2X-Large
19-555-472 Hi-vis Lime 3X-Large
19-555-475 Hi-vis Orange Medium
19-555-474 Hi-vis Orange Large
19-555-478 Hi-vis Orange X-Large
19-555-476 Hi-vis Orange 2X-Large
19-555-477 Hi-vis Orange 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-555-469 Supplier Pyramex™ Supplier No. RVHL2910M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Hi-Vis Reflective Safety Vests with ANSI/ISEA 107-2015 safety compliance
  • Front: Hook & Loop closure
  • One 2 in. silver reflective stripe applied horizontally 8.5 in. from the bottom
  • Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over shoulders to top of back side horizontal stripe
Specifications
Closure Type Hook and Loop
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 2
Color Hi-vis Lime
Size Medium
Material Polyester
Size (English) Chest 38 to 40 in.
Tape Type China Brand Flat Silver