Learn More
Pyramex™ Hi-Vis Reflective T-Shirts
Catalog No. 19555425 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
2X-Large
3X-Large
4X-Large/5X-Large
Large
Medium
X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-425 Hi-vis Orange 4X-Large/5X-Large
19-555-418 Hi-vis Lime 3X-Large
19-555-419 Hi-vis Lime 4X-Large/5X-Large
19-555-422 Hi-vis Orange Medium
19-555-423 Hi-vis Orange 2X-Large
19-555-416 Hi-vis Lime Medium
19-555-415 Hi-vis Lime Large
19-555-420 Hi-vis Lime X-Large
19-555-417 Hi-vis Lime 2X-Large
19-555-421 Hi-vis Orange Large
19-555-426 Hi-vis Orange X-Large
19-555-424 Hi-vis Orange 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-555-425 Supplier Pyramex™ Supplier No. RTS3320X4X5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Hi-Vis Reflective T-Shirts with ANSI/ISEA 107-2015 safety compliance
  • One 2 in. silver reflective stripe applied horizontally across lower chest
  • One 2 in. silver reflective stripe applied around each sleeve
  • Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over shoulders to top of back side horizontal stripe
  • Outside left chest pocket measures 5.5 x 5.5 in.
Specifications
Size 4X-Large/5X-Large
Material Polyester
Color Hi-vis Orange
Sleeve Style Short Sleeve
No. of Pockets 1
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 3
Size (English) Chest 58 to 64 in.
Tape Type China Brand Flat Silver