Learn More
MSC Mcr Safety Luminator Safety Vest, Polyester, Lime, 5XL, 1 Ea.
Catalog No. 5000686405
Encompass
This high-visibility safety vest is engineered for workers in roadway and temporary traffic control zones. Constructed with a solid front and mesh back, the garment provides comfort and breathability in high-traffic environments while meeting safety requirements.
  • Complies with ANSI/ISEA 107 2020 Type R Class 2 standards
  • Solid front and mesh back construction
  • Easy release zipper front closure
  • Features 4 in. orange and 2 in. silver reflective striping
  • Includes three pockets with a clear ID pocket on the right chest
  • Integrated mic holder tab on the left shoulder
  • Five-point breakaway design
Catalog No. 50-006-86405 Supplier MSC Supplier No. 34064584
This high-visibility safety vest is engineered for workers in roadway and temporary traffic control zones. Constructed with a solid front and mesh back, the garment provides comfort and breathability in high-traffic environments while meeting safety requirements.
  • Complies with ANSI/ISEA 107 2020 Type R Class 2 standards
  • Solid front and mesh back construction
  • Easy release zipper front closure
  • Features 4 in. orange and 2 in. silver reflective striping
  • Includes three pockets with a clear ID pocket on the right chest
  • Integrated mic holder tab on the left shoulder
  • Five-point breakaway design