Learn More
MSC Tingley Profile PVC Knee Boot, Plain Toe, Dark Brown, 1 ea
Catalog No. 5000686577
Encompass
These plain toe knee boots are designed for performance and value in agricultural, industrial, and food processing applications. Constructed using high-quality PVC resins and specialized low-temperature additives, they maintain excellent flexibility and durability in cold environments.
  • Injection-molded seamless construction provides 100% liquidproof protection
  • Flexible upper material remains supple in cold temperatures
  • Cleated outsole design expels debris to maintain traction on various surfaces
  • Calf Relief Expansion™ feature allows for an additional 1.5 in. of calf comfort
  • Removable three-layer polyurethane contour insole wicks perspiration and absorbs shock
  • Beveled heel reduces leg and back strain
  • Slip resistant per ASTM F3445 standards
  • Integrated tread wear indicators
Catalog No. 50-006-86577 Supplier MSC Supplier No. 46017521
These plain toe knee boots are designed for performance and value in agricultural, industrial, and food processing applications. Constructed using high-quality PVC resins and specialized low-temperature additives, they maintain excellent flexibility and durability in cold environments.
  • Injection-molded seamless construction provides 100% liquidproof protection
  • Flexible upper material remains supple in cold temperatures
  • Cleated outsole design expels debris to maintain traction on various surfaces
  • Calf Relief Expansion™ feature allows for an additional 1.5 in. of calf comfort
  • Removable three-layer polyurethane contour insole wicks perspiration and absorbs shock
  • Beveled heel reduces leg and back strain
  • Slip resistant per ASTM F3445 standards
  • Integrated tread wear indicators