Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Hi-Vis Synthetic Knit Palm Coated Gloves

Catalog No. 19148853 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
2X-Large
Large
Medium
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-853 2X-Large
19-148-852 X-Large
19-148-851 Large
19-148-850 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-148-853 Supplier Wells Lamont™ Supplier No. Y9239TXXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide additional protection in high wear/high risk areas of hand. FlexTech™ Hi-Vis Synthetic Knit Palm Coated Gloves are light weight gloves that offer great awareness in the workplace.

  • Can be used in temperatures from -70°F to 300°F
  • Can handle dry ice for short periods of time
  • Hi-Vis green light weight synthetic knit shell
  • Latex palm provides outstanding grip
  • Knit wrist construction prevents dirt and debris from directly contacting the hand
  • Non-slip surface allows workers to work more efficiently and increase their grip
  • The open, uncoated back circulates air, allowing hands to breathe and keeping them cool and comfortable
  • ANSI Level A3 cut resistance

Specifications

Coating Material Latex
Color Lime Green
Cut Resistance Rating ANSI A3
Latex Free No
Powdered Powder Free
Product Type Synthetic Knit Shell
Size 2X-Large
Sterility Non-sterile
Temperature (English) Max. 300°F
Temperature (Metric) Max 148.89°C
For Use With (Application) Cold Temperatures
Puncture Rating ANSI/ISEA 105-16 Level 1
Tear Rating 3
Temperature (English) Min -70°F
Temperature (Metric) Min. -56.67°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.