Learn More
Pyramex Safety Products Hi-Visibility Self-Extinguishing Safety Vest
Catalog No. 19555155 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Medium
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-155 Lime Medium
19-555-156 Lime 2X-Large
19-555-158 Orange Medium
19-555-159 Orange 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-555-155 Supplier Pyramex Safety Products Supplier No. RCA2510SEM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex Safety™ Hi-Visibility Self-Extinguishing Safety Vest features 2 in. silver reflective material.
  • Meets ANSI/ISEA 107-2015 and CSA Z96-15 standards
  • Hi-Vis, self-extinguishing, lightweight polyester mesh material.
  • Breakaway hook and loop shoulders, sides, and front closure
  • Adjustable waist band
  • 3 pockets: 1 outside, 2 inside
  • Outside chest pocket with hook and loop closure
  • Deep and wide inside pockets
  • Reinforced D-ring slot
Specifications
Certifications/Compliance ANSI/ISEA 107-2015; CSA Z96-15
Color Lime
Size Medium
Material Polyester
No. of Pockets 3
Tape Type 2 in. Silver reflective stripes