Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ M-PACT™ Impact Resistant Gloves, Hi-Viz, with D30™ Padding

Catalog No. 19030111 Shop All Mechanix Wear Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Large
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-030-111 Medium
19-030-112 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-030-111 Supplier Mechanix Wear™ Supplier No. SMP91009
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Hi-Viz M-PACT protects working hands with Thermoplastic Rubber (TPR) knuckle protection and eye-catching luminance in low-light environments. Meets EN 388 impact standard to help prevent blunt force impact injuries in the field.

  • Thermoplastic Rubber (TPR) pinch-point fingertip protection
  • High-visibility TrekDry™ keeps hands cool and comfortable
  • Reflective print provides eye-catching luminance in low-light environments
  • D3O™ palm padding absorbs and dissipates high-impact energy
  • Armortex™ palm reinforcement provides added durability
  • Reinforced index finger and thumb deliver durability where its needed most
  • Hook and Loop closure provides a secure fit
  • Touchscreen capable synthetic leather to stay connected while on the job
  • Machine washable

Specifications

ANSI Abrasion Rating ANSI ISEA 138 (level 2), EN 388
Back Material TrekDry™
Cuff Material TPR
Color Fluorescent Yellow
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size Medium
Finger Style Full
For Use With (Application) Construction, Heavy Equipment Operation, Towing, and Transportation
Model SMP-91
Series M-Pact™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.