Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Hi3 Ecoli Standard, 6 Synthetically Prepared Peptides, Max Recovery Vial

Catalog No. 50466281 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Hi3 Ecoli Standard is a set of 6 highly purified peptides, offering the top 6 ionizing peptides of the E. coli ClpB protein. Quantitated via AAA to equimolar ratios and verified over 95% pure, these standards are designed for microbial samples and the Hi3 quantification method in MSE proteomics data.

  • Set of 6 highly purified peptides.
  • Top 6 ionizing peptides of E. coli ClpB protein.
  • Quantitated via AAA to equimolar ratios.
  • Over 95% purity, mass identification verified.
  • Packaged in a Max Recovery vial.
  • Contains 1 nmol of each peptide per vial.
  • Reconstitution: 100 μL of 0.5% trifluoroacetic acid in 3% acetonitrile/water for 10 pmol/μL stock.
  • For Hi3 quantification method in MSE proteomics.
  • Suitable for LC/MS system evaluation and benchmarking.
  • Non-hazardous peptides, powder form, laboratory use only.

Catalog No. 50-466-281 Supplier Waters Corp Supplier No. 186006012
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Hi3 Ecoli Standard is a set of 6 highly purified peptides, offering the top 6 ionizing peptides of the E. coli ClpB protein. Quantitated via AAA to equimolar ratios and verified over 95% pure, these standards are designed for microbial samples and the Hi3 quantification method in MSE proteomics data.

  • Set of 6 highly purified peptides.
  • Top 6 ionizing peptides of E. coli ClpB protein.
  • Quantitated via AAA to equimolar ratios.
  • Over 95% purity, mass identification verified.
  • Packaged in a Max Recovery vial.
  • Contains 1 nmol of each peptide per vial.
  • Reconstitution: 100 μL of 0.5% trifluoroacetic acid in 3% acetonitrile/water for 10 pmol/μL stock.
  • For Hi3 quantification method in MSE proteomics.
  • Suitable for LC/MS system evaluation and benchmarking.
  • Non-hazardous peptides, powder form, laboratory use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.