Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Standard, Hi3 Phos B, Solid, Peptide

Catalog No. 50466280 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Hi3 Phos B Standard comprises six synthetically prepared, highly purified peptides, quantified via AAA for enhanced accuracy and over 95% purity. Intended for Hi3 quantification of MSE proteomics data from microbial samples, it also evaluates and benchmarks proteomic LC/MS systems.

  • Six synthetically prepared, highly purified peptides.
  • Quantified via AAA to equimolar ratios; >95% pure.
  • For Hi3 quantification of MSE proteomics data.
  • Suitable for microbial samples and LC/MS system benchmarking.
  • Shelf life: 36 months (unreconstituted), 1 week (reconstituted).
  • Storage: −20°C (medium–term); shipped ambient.
  • Reconstitution: 100 μL of 0.5% trifluoroacetic acid in 3% acetonitrile/water.

Catalog No. 50-466-280 Supplier Waters Corp Supplier No. 186006011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Hi3 Phos B Standard comprises six synthetically prepared, highly purified peptides, quantified via AAA for enhanced accuracy and over 95% purity. Intended for Hi3 quantification of MSE proteomics data from microbial samples, it also evaluates and benchmarks proteomic LC/MS systems.

  • Six synthetically prepared, highly purified peptides.
  • Quantified via AAA to equimolar ratios; >95% pure.
  • For Hi3 quantification of MSE proteomics data.
  • Suitable for microbial samples and LC/MS system benchmarking.
  • Shelf life: 36 months (unreconstituted), 1 week (reconstituted).
  • Storage: −20°C (medium–term); shipped ambient.
  • Reconstitution: 100 μL of 0.5% trifluoroacetic acid in 3% acetonitrile/water.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.