Learn More
HANNA INSTRUMENTS INC pH/mV Simulator (w/ screen)
SDP
Catalog No. NC1773859
Encompass
HI931001 is a portable pH/mV simulators designed specifically for calibrating pH and mV (ORP) meters.Simulate output to discover malfunctions.Universal BNC connector.Battery power allows for use away from a power source.
Catalog No. NC1773859 Supplier HANNA INSTRUMENTS INC Supplier No. HI931001
HI931001 is a portable pH/mV simulators designed specifically for calibrating pH and mV (ORP) meters.Simulate output to discover malfunctions.Universal BNC connector.Battery power allows for use away from a power source.