Learn More
Aviva Systems Biology Corp HIBADH Antibody - middle region (ARP53112_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53112_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIBADH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIBADH. Alternate names: MGC40361, NS5ATP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60926