Learn More
BrainBits Hibernate A, 100 mL
Catalog No. NC1787837
Encompass
Hibernate is a nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels.
Catalog No. NC1787837 Supplier BrainBits Supplier No. HA100
Hibernate is a nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels.