Learn More
BrainBits Hibernate A Low-Fluorescence, 500ml
Catalog No. NC0442869
Encompass
Hibernate is a nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels (US Patent 6,180,404).Transnetyx Tissue is the sole supplier of custom Hibernate Medium.Hibernate-E is useful for embryonic tissue while Hibernate-A is useful for adult tissue.
Catalog No. NC0442869 Supplier BrainBits Supplier No. HALF500
Hibernate is a nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels (US Patent 6,180,404).Transnetyx Tissue is the sole supplier of custom Hibernate Medium.Hibernate-E is useful for embryonic tissue while Hibernate-A is useful for adult tissue.