Learn More
BrainBits HIBERNATE E 500ML
Catalog No. NC9063748
Encompass
Nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels
Catalog No. NC9063748 Supplier BrainBits Supplier No. HE500
Nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels