Learn More
Molnlycke Hibiclens™ Skin Cleanser
Catalog No. 22355135
Change view
Click to view available options
Volume (English):
4 oz.
8 oz.
16 oz.
32 oz.
1 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-355-135 16 oz.
22-355-133 4 oz.
22-355-134 8 oz.
22-355-136 32 oz.
22-355-137 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-355-135 Supplier Molnlycke Supplier No. MOL 57516
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hibiclens has been trusted by healthcare professionals for more than 40 years. Its efficacy and safety is proven. This antiseptic skin cleanser provides effective defense against the spread of infections. The active ingredient, chlorhexidine gluconate, begins to kill germs on contact.
  • With multiple indications, Hibiclens has a great deal of utility throughout the hospital facility. The 4% chlorhexidine gluconate solution is in a gentle, skin cleansing base that can be used for:
    • Daily patient bed baths, as part of a horizontal infection prevention approach
    • Preoperative/postoperative showering and decolonization, as part of surgical site infection prevention bundles
    • Surgical hand scrubbing
    • Personnel hand washing
    • Preoperative skin preparation
Specifications
Product Type Antiseptic Skin Cleanser
Form Liquid
For Use With (Application) Daily Patient Bed Baths, Preoperative/Postoperative Showering, Surgical Hand Scrubbing, Personnel Hand Washing, Preoperative Skin Preparation
Quantity 12/Cs.
Volume (English) 16 oz.