Learn More
Aviva Systems Biology Corp HIC1 Antibody - middle region (ARP38806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIC1. Alternate names: ZBTB29, hic-1, ZNF901ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56808