Learn More
Aviva Systems Biology Corp HIC1 Antibody - N-terminal region (ARP32623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIon Channel1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HIC1. Alternate names: hic-1, ZBTB29, ZNF901ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54678