Learn More
Aviva Systems Biology Corp HIC2 Antibody - middle region (ARP32981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIC2. Alternate names: HRG22, KIAA1020, ZBTB30, ZNF907ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54841