Learn More
Strem, An Ascensus Company CAS 403815-19-0. 100mg. 44-Bis(t-butyl)-1122-tetrakis(diphenylphosphino)ferrocene 98% HiersoPHOS-5. MFCD09971735
SDP
Supplier:  Strem, An Ascensus Company 260326100MG
Encompass
CAS 403815-19-0. 100mg. 44-Bis(t-butyl)-1122-tetrakis(diphenylphosphino)ferrocene 98% HiersoPHOS-5. MFCD09971735. Molecular Weight 1034.94. Molecular Formula C66H62FeP4. Color/form orange xtl. Strem 26-0326. http//www.strem.com/catalog/v/26-0326/
Catalog No. 50-211-0793