Learn More
Strem, An Ascensus Company CAS 109313-83-9. 500mg. 11-Bis(1-diphenylphosphino-1-methylethyl)ferrocene ethanol adduct 97% HiersoPHOS-6 (Sylphos). MFCD28144554
SDP
Supplier:  Strem, An Ascensus Company 260290500MG
Encompass
CAS 109313-83-9. 500mg. 11-Bis(1-diphenylphosphino-1-methylethyl)ferrocene ethanol adduct 97% HiersoPHOS-6 (Sylphos). MFCD28144554. Molecular Weight 638.54 (684.61). Molecular Formula C40H40FeP2CH3CH2OH. Color/form orange solid Strem 26-0290. http//www.strem.com/catalog/v/26-0290/
Catalog No. 50-211-0792