Learn More
Enzo Life Sciences HIF-1 alpha monoclonal antibody (H1alpha67) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2870100
Encompass_Preferred
Clone: H1α67. Isotype: IgG. Immunogen: Recombinant human hypoxia-inducible factor-1α (HIF-1α) (aa 432-528). Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Ferret, Bovine. Applications: ELISA, Flow Cytometry, ICC, IF, IHC, IHC (FS), IHC (PS), IP, WB. Purity: Purified IgG. Formulation: Liquid. In PBS with 1% BSA containing 0.05% sodium azide.
Catalog No. 50-201-1792