Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HIF-1α-IN-2 hydrochloride | 00-00-0 | 98.5% | 397.92 g·mol⁻¹ | C21H20ClN3OS | 5 MG
SDP

Catalog No. 5000264508
Encompass_Preferred

HIF-1α-IN-2 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of hypoxia-inducible factor 1-alpha (HIF-1α) used in research to probe HIF-1α-related signaling and anticancer activity. The compound is supplied as a powdered research reagent with reported cellular potency and a certificate of analysis for identity and purity.

  • Inhibits HIF-1α with cellular IC50s of ~28 nM (MDA-MB-231) and ~15 nM (MiaPaCa-2).
  • Molecular formula C21H20ClN3OS; molecular weight 397.92 g·mol⁻¹.
  • Purity 98.5% (measured by LCMS on the COA).
  • Provided as the hydrochloride salt for research use.
  • Available in small milligram pack sizes suitable for laboratory assays.

Catalog No. 50-002-64508 Supplier Medchemexpress LLC Supplier No. HY115903A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HIF-1α-IN-2 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of hypoxia-inducible factor 1-alpha (HIF-1α) used in research to probe HIF-1α-related signaling and anticancer activity. The compound is supplied as a powdered research reagent with reported cellular potency and a certificate of analysis for identity and purity.

  • Inhibits HIF-1α with cellular IC50s of ~28 nM (MDA-MB-231) and ~15 nM (MiaPaCa-2).
  • Molecular formula C21H20ClN3OS; molecular weight 397.92 g·mol⁻¹.
  • Purity 98.5% (measured by LCMS on the COA).
  • Provided as the hydrochloride salt for research use.
  • Available in small milligram pack sizes suitable for laboratory assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.